siRNA Details
| Gene Symbol: 9530026F06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040483.1 | 2 | 212 | |||
| GenBank mRNA | AK020574.1 | - | 2 | 213 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9530026F06Rik| Gene Symbol: 9530026F06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040483.1 | 2 | 212 | |||
| GenBank mRNA | AK020574.1 | - | 2 | 213 | ||