siRNA Details
| Gene Symbol: 1110046J04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040707.1 | 4 | 457 | |||
| GenBank mRNA | AK004186.1 | - | 4 | 457 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110046J04Rik| Gene Symbol: 1110046J04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040707.1 | 4 | 457 | |||
| GenBank mRNA | AK004186.1 | - | 4 | 457 | ||