siRNA Details
| Gene Symbol: 4930527E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019703.1 | - | 1, 2 | 185 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930527E20Rik| Gene Symbol: 4930527E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019703.1 | - | 1, 2 | 185 | ||