siRNA Details
| Gene Symbol: 4930517E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040611.1 | 3 | 1307 | |||
| GenBank mRNA | AK015809.1 | - | 3 | 1307 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930517E11Rik| Gene Symbol: 4930517E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040611.1 | 3 | 1307 | |||
| GenBank mRNA | AK015809.1 | - | 3 | 1307 | ||