siRNA Details
| Gene Symbol: Gm12320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039373.1 | - | 4 | 575 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB633298.1 | - | 4 | 575 | Chr. 11 - 70749253 - 70755151 [-] on Build GRCm38 | |

Gm12320
Genomic Map