siRNA Details
| Gene Symbol: C630028M04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040668.1 | 3 | 2470 | |||
| GenBank mRNA | AK083215.1 | - | 3 | 2471 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C630028M04Rik| Gene Symbol: C630028M04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040668.1 | 3 | 2470 | |||
| GenBank mRNA | AK083215.1 | - | 3 | 2471 | ||