siRNA Details
Gene Symbol: C630028M04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040668.1 | 3 | 1048 | |||
GenBank mRNA | AK083215.1 | - | 3 | 1049 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C630028M04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040668.1 | 3 | 1048 | |||
GenBank mRNA | AK083215.1 | - | 3 | 1049 |