siRNA Details
| Gene Symbol: 1810063I02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007947.1 | - | 2 | 187 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810063I02Rik| Gene Symbol: 1810063I02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007947.1 | - | 2 | 187 | ||