siRNA Details
| Gene Symbol: 2610028E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015560.1 | 1 | 223 | |||
| GenBank mRNA | AK031953.1 | - | 1 | 223 | ||
| AK083757.1 | - | 1 | 317 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY718444.1 | - | 1 | 223 | Chr. 4 - 125891482 - 125922310 [-] on Build GRCm38 | |

2610028E06Rik
Genomic Map