siRNA Details
| Gene Symbol: Gm12602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144841.1 | - | 4 | 544 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY734487.1 | - | 4 | 544 | Chr. 4 - 89100664 - 89128226 [-] on Build GRCm38 | |

Gm12602
Genomic Map