siRNA Details
| Gene Symbol: 4930589P08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016376.1 | - | 2 | 374 | ||
| AK017048.1 | - | 2 | 368 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930589P08Rik| Gene Symbol: 4930589P08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016376.1 | - | 2 | 374 | ||
| AK017048.1 | - | 2 | 368 | |||