siRNA Details
| Gene Symbol: Gm16041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044237.1 | - | 1 | 62 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB591428.2 | - | 1 | 62 | Chr. 1 - 4970857 - 4973744 [+] on Build GRCm38 | |

Gm16041
Genomic Map