siRNA Details
| Gene Symbol: 4930465M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045973.1 | 2 | 305 | |||
| GenBank mRNA | AK015269.1 | - | 2 | 328 | ||
| AK015510.1 | - | 2 | 305 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930465M20Rik| Gene Symbol: 4930465M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045973.1 | 2 | 305 | |||
| GenBank mRNA | AK015269.1 | - | 2 | 328 | ||
| AK015510.1 | - | 2 | 305 | |||