siRNA Details
| Gene Symbol: Gm12992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102394.1 | 3 | 712 | |||
| GenBank mRNA | AK047689.1 | - | 2 | 446 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB647646.1 | - | 2 | 443 | Chr. 4 - 131913201 - 131915514 [-] on Build GRCm38 | |

Gm12992
Genomic Map