siRNA Details
| Gene Symbol: Mettl5os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016085.1 | - | 1 | 237 | ||
| AK050748.1 | - | 1 | 306 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB654546.1 | - | 1 | 304 | Chr. 2 - 69885660 - 69886268 [+] on Build GRCm38 | |

Mettl5os
Genomic Map