siRNA Details
| Gene Symbol: Lrrc75aos2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005754.1 | - | 2 | 421 | ||
| AK015368.1 | - | 2 | 479 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc75aos2| Gene Symbol: Lrrc75aos2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005754.1 | - | 2 | 421 | ||
| AK015368.1 | - | 2 | 479 | |||