siRNA Details
| Gene Symbol: Smc2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045175.1 | 2 | 517 | |||
| GenBank mRNA | AK012547.1 | - | 2 | 353 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smc2os| Gene Symbol: Smc2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045175.1 | 2 | 517 | |||
| GenBank mRNA | AK012547.1 | - | 2 | 353 | ||