siRNA Details
| Gene Symbol: Gm15624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038985.1 | - | 3 | 1050 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK137106.1 | - | 2600 | Chr. - - [-] on Build | ||

Gm15624
Genomic Map