siRNA Details
| Gene Symbol: Gm14718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038463.1 | 2 | 559 | |||
| GenBank mRNA | AK041463.1 | - | 2 | 507 | ||
| BC039994.1 | - | 2 | 553 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14718| Gene Symbol: Gm14718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038463.1 | 2 | 559 | |||
| GenBank mRNA | AK041463.1 | - | 2 | 507 | ||
| BC039994.1 | - | 2 | 553 | |||