siRNA Details
| Gene Symbol: Ube4bos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020898.1 | - | 443 | |||
| BC024827.1 | - | 2 | 413 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube4bos1| Gene Symbol: Ube4bos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020898.1 | - | 443 | |||
| BC024827.1 | - | 2 | 413 | |||