siRNA Details
| Gene Symbol: 4933406K04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015512.2 | 3, 4 | 437 | |||
| GenBank mRNA | AK015208.1 | - | 3, 4 | 438 | ||
| AK015628.1 | - | 3, 4 | 447 | |||
| AK016698.1 | - | 3, 4 | 477 | |||
| AK016950.1 | - | 2, 3 | 332 | |||

4933406K04Rik
Genomic Map