siRNA Details
| Gene Symbol: 4933406K04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015512.2 | 3 | 401 | |||
| GenBank mRNA | AK015208.1 | - | 3 | 402 | ||
| AK015628.1 | - | 3 | 411 | |||
| AK016698.1 | - | 3 | 441 | |||
| AK016950.1 | - | 2 | 296 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933406K04Rik| Gene Symbol: 4933406K04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015512.2 | 3 | 401 | |||
| GenBank mRNA | AK015208.1 | - | 3 | 402 | ||
| AK015628.1 | - | 3 | 411 | |||
| AK016698.1 | - | 3 | 441 | |||
| AK016950.1 | - | 2 | 296 | |||