siRNA Details
| Gene Symbol: 2410114N07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040652.1 | 3 | 256 | |||
| GenBank mRNA | AK010767.1 | - | 3 | 257 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY710199.1 | - | 257 | Chr. - - [-] on Build | ||

2410114N07Rik
Genomic Map