siRNA Details
| Gene Symbol: 2410003L11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045496.1 | 4 | 590 | |||
| GenBank mRNA | AK010368.1 | - | 4 | 591 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410003L11Rik| Gene Symbol: 2410003L11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045496.1 | 4 | 590 | |||
| GenBank mRNA | AK010368.1 | - | 4 | 591 | ||