siRNA Details
| Gene Symbol: Gm13483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040362.1 | 3 | 446 | |||
| GenBank mRNA | AK045825.1 | - | 3 | 392 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB644362.1 | - | 3 | 392 | Chr. 2 - 50296865 - 50433981 [+] on Build GRCm38 | |

Gm13483
Genomic Map