siRNA Details
| Gene Symbol: Gm12409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046068.1 | 3 | 289 | |||
| GenBank mRNA | AK041524.1 | - | 3 | 290 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB637831.1 | - | 3 | 286 | Chr. 4 - 45762375 - 45764220 [-] on Build GRCm38 | |

Gm12409
Genomic Map