siRNA Details
| Gene Symbol: 1700054M17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045919.1 | 3 | 370 | |||
| GenBank mRNA | AK006785.1 | - | 3 | 370 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706771.1 | - | 3 | 370 | Chr. 2 - 118304914 - 118308166 [+] on Build GRCm38 | |

1700054M17Rik
Genomic Map