siRNA Details
| Gene Symbol: A930041C12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046195.1 | 3 | 394 | |||
| GenBank mRNA | AK020948.1 | - | 3 | 395 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930041C12Rik| Gene Symbol: A930041C12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046195.1 | 3 | 394 | |||
| GenBank mRNA | AK020948.1 | - | 3 | 395 | ||