siRNA Details
| Gene Symbol: 4930405D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045956.1 | 3 | 332 | |||
| GenBank mRNA | AK015094.1 | - | 2 | 311 | ||
| AK147079.1 | - | 3 | 333 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930405D11Rik| Gene Symbol: 4930405D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045956.1 | 3 | 332 | |||
| GenBank mRNA | AK015094.1 | - | 2 | 311 | ||
| AK147079.1 | - | 3 | 333 | |||