siRNA Details
| Gene Symbol: 2310016D23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009383.1 | - | 4 | 367 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310016D23Rik| Gene Symbol: 2310016D23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009383.1 | - | 4 | 367 | ||