siRNA Details
| Gene Symbol: 1110028F11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045139.1 | 2 | 2434 | |||
| GenBank mRNA | AK003943.1 | - | 2 | 563 | ||
| BY594060.1 | - | 1 | 206 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110028F11Rik| Gene Symbol: 1110028F11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045139.1 | 2 | 2434 | |||
| GenBank mRNA | AK003943.1 | - | 2 | 563 | ||
| BY594060.1 | - | 1 | 206 | |||