siRNA Details
Gene Symbol: 9530051G07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040272.1 | 4 | 3505 | |||
GenBank mRNA | AK032014.1 | - | 3 | 3206 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9530051G07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040272.1 | 4 | 3505 | |||
GenBank mRNA | AK032014.1 | - | 3 | 3206 |