siRNA Details
| Gene Symbol: 1700001G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038077.1 | 1 | 94 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF104711.1 | - | 1 | 94 | Chr. 14 - 66295516 - 66297129 [-] on Build GRCm38 | |

1700001G11Rik
Genomic Map