siRNA Details
| Gene Symbol: A830012C17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043612.1 | - | 1349 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A830012C17Rik| Gene Symbol: A830012C17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043612.1 | - | 1349 | |||