siRNA Details
| Gene Symbol: Gm15706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045598.1 | 2 | 638 | |||
| GenBank mRNA | AK084826.1 | - | 2 | 639 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15706| Gene Symbol: Gm15706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045598.1 | 2 | 638 | |||
| GenBank mRNA | AK084826.1 | - | 2 | 639 | ||