siRNA Details
| Gene Symbol: Gm12374 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029761.1 | - | 405 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615591.1 | - | 4 | 405 | Chr. 4 - 37249403 - 37320412 [+] on Build GRCm38 | |

Gm12374
Genomic Map