siRNA Details
| Gene Symbol: 1700095A21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045468.1 | 3 | 290 | |||
| GenBank mRNA | AK007074.1 | - | 3 | 291 | ||
| AK007118.1 | - | 3 | 292 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700095A21Rik| Gene Symbol: 1700095A21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045468.1 | 3 | 290 | |||
| GenBank mRNA | AK007074.1 | - | 3 | 291 | ||
| AK007118.1 | - | 3 | 292 | |||