siRNA Details
| Gene Symbol: Unc45bos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040863.1 | - | 2 | 463 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc45bos| Gene Symbol: Unc45bos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040863.1 | - | 2 | 463 | ||