siRNA Details
| Gene Symbol: 5730457N03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038163.1 | 4 | 2426 | |||
| GenBank mRNA | AK017671.1 | - | 3 | 2299 | ||
| AK031498.1 | - | 4 | 2428 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5730457N03Rik| Gene Symbol: 5730457N03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038163.1 | 4 | 2426 | |||
| GenBank mRNA | AK017671.1 | - | 3 | 2299 | ||
| AK031498.1 | - | 4 | 2428 | |||