siRNA Details
| Gene Symbol: St8sia3os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007031.1 | - | 3 | 297 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707098.1 | - | 3 | 297 | Chr. 18 - 64200679 - 64277180 [-] on Build GRCm38 | |

St8sia3os
Genomic Map