siRNA Details
| Gene Symbol: Ccdc34os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040508.1 | 5 | 772 | |||
| GenBank mRNA | AK006310.1 | - | 5 | 773 | ||
| BC048603.1 | - | 2 | 442 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc34os| Gene Symbol: Ccdc34os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040508.1 | 5 | 772 | |||
| GenBank mRNA | AK006310.1 | - | 5 | 773 | ||
| BC048603.1 | - | 2 | 442 | |||