siRNA Details
| Gene Symbol: Zfp652os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045780.1 | 2 | 242 | |||
| GenBank mRNA | AK027957.1 | - | 2 | 243 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp652os| Gene Symbol: Zfp652os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045780.1 | 2 | 242 | |||
| GenBank mRNA | AK027957.1 | - | 2 | 243 | ||