siRNA Details
Gene Symbol: 4933405E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045506.1 | 3 | 941 | |||
GenBank mRNA | AK016664.1 | - | 3 | 944 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933405E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045506.1 | 3 | 941 | |||
GenBank mRNA | AK016664.1 | - | 3 | 944 |