siRNA Details
| Gene Symbol: Srrm4os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015595.2 | 4 | 876 | |||
| GenBank mRNA | AK049160.1 | - | 4 | 877 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF463123.1 | - | 1 | 333 | Chr. 5 - 116462676 - 116463161 [+] on Build GRCm38 | |

Srrm4os
Genomic Map