siRNA Details
| Gene Symbol: 2700069I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045905.2 | 3 | 329 | |||
| GenBank mRNA | AK012506.1 | - | 3 | 332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2700069I18Rik| Gene Symbol: 2700069I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045905.2 | 3 | 329 | |||
| GenBank mRNA | AK012506.1 | - | 3 | 332 | ||