siRNA Details
| Gene Symbol: 4930483K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045354.1 | 4 | 785 | |||
| GenBank mRNA | AK015616.1 | - | 4 | 787 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930483K19Rik| Gene Symbol: 4930483K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045354.1 | 4 | 785 | |||
| GenBank mRNA | AK015616.1 | - | 4 | 787 | ||