siRNA Details
Gene Symbol: 4930483K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045354.1 | 4 | 746 | |||
GenBank mRNA | AK015616.1 | - | 4 | 748 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930483K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045354.1 | 4 | 746 | |||
GenBank mRNA | AK015616.1 | - | 4 | 748 |