siRNA Details
| Gene Symbol: 3100003L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045907.1 | 3 | 457 | |||
| GenBank mRNA | AK013926.1 | - | 3 | 457 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3100003L05Rik| Gene Symbol: 3100003L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045907.1 | 3 | 457 | |||
| GenBank mRNA | AK013926.1 | - | 3 | 457 | ||