siRNA Details
| Gene Symbol: Rbm46os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040381.1 | 4 | 2704 | |||
| NR_040382.1 | 4 | 2695 | ||||
| GenBank mRNA | AK015378.1 | - | 919 | |||
| AK161442.1 | - | 4 | 2705 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm46os| Gene Symbol: Rbm46os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040381.1 | 4 | 2704 | |||
| NR_040382.1 | 4 | 2695 | ||||
| GenBank mRNA | AK015378.1 | - | 919 | |||
| AK161442.1 | - | 4 | 2705 | |||