siRNA Details
Gene Symbol: Rbm46os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040381.1 | 4 | 1468 | |||
NR_040382.1 | 4 | 1459 | ||||
GenBank mRNA | AK161442.1 | - | 4 | 1469 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rbm46os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040381.1 | 4 | 1468 | |||
NR_040382.1 | 4 | 1459 | ||||
GenBank mRNA | AK161442.1 | - | 4 | 1469 |